Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP8B1 Antibody (M15-P3B7), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250218F
Description
CYP8B1 Monoclonal specifically detects CYP8B1 in Human, Rat samples. It is validated for ELISA, Immunohistochemistry.Specifications
CYP8B1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
CYP8B1 | |
Peptide sequence- QPSHDVRFR (amino acids 486 - 494) | |
0.1 mL | |
Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
1582 | |
Store at 4C in the dark. | |
IgG1 κ |
ELISA, Immunohistochemistry | |
M15-P3B7 | |
ELISA, Immunohistochemistry | |
CP8B, CYP127 alpha-hydroxy-4-cholesten-3-one 12-alpha-hydroxylase, CYPVIIIB1, Cytochrome P450 8B1, cytochrome P450, family 8, subfamily B, polypeptide 1, cytochrome P450, subfamily VIIIB (sterol 12-alpha-hydroxylase), polypeptide 1,7-alpha-hydroxycholest-4-en-3-one 12-alpha-hydroxylase, EC 1.14.13.95, FLJ17826,7-alpha-hydroxy-4-cholesten-3-one 12-alpha-hydroxylase, Sterol 12-alpha-hydroxylase | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction