Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cypermethrin, 98%, ACROS Organics™
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 1.1200g/mL |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (Mixture of isomers) (HPLC) |
Quantity | 5 g |
Specific Gravity | 1.12 |
Solubility Information | Solubility in water: insoluble. |
Formula Weight | 416.3 |
Percent Purity | 96% |
Chemical Name or Material | Cypermethrin |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Toxic if inhaled. Toxic if swallowed. May cause respiratory irritation. Very toxic to aquatic life with long lasting effects. |
Precautionary Statement | GHS P Statement Wash face,hands and any exposed skin thoroughly after handling. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: Remove to fresh air and |
EINECSNumber | 257-842-9 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction