Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CyPPA, Tocris Bioscience™

Click to view available options
Quantity:
10mg
This item is not returnable. View return policy
73029-73-9
C16H23N5
285.395
USEMRPYUFJNFQN-UHFFFAOYSA-N
cyppa,n-cyclohexyl-2-3,5-dimethyl-1h-pyrazol-1-yl-6-methylpyrimidin-4-amine,cyclohexyl-2-3,5-dimethyl-pyrazol-1-yl-6-methyl-pyrimidin-4-yl-amine,n-cyclohexyl-2-3,5-dimethylpyrazol-1-yl-6-methylpyrimidin-4-amine,n-cyclohexyl-n-2-3,5-dimethyl-pyrazol-1-yl-6-methyl-4-pyrimidinamine,d00dif,cyppa hplc , solid,2-3,5-dimethyl-1h-pyrazole-1-yl-4-methyl-6-cyclohexylamino pyrimidine,n-cyclohexyl-2-3,5-dimethyl-1h-pyrazol-1-yl-6-methyl-4-pyrimidinamine,n-cyclohexyl-n-2-3,5-dimethyl-pyrazol-1-yl-6-methyl-4-pyrimidinamine
909822
N-cyclohexyl-2-(3,5-dimethylpyrazol-1-yl)-6-methylpyrimidin-4-amine
CC1=CC(=NC(=N1)N2C(=CC(=N2)C)C)NC3CCCCC3
This item is not returnable. View return policy
73029-73-9
C16H23N5
285.395
USEMRPYUFJNFQN-UHFFFAOYSA-N
cyppa,n-cyclohexyl-2-3,5-dimethyl-1h-pyrazol-1-yl-6-methylpyrimidin-4-amine,cyclohexyl-2-3,5-dimethyl-pyrazol-1-yl-6-methyl-pyrimidin-4-yl-amine,n-cyclohexyl-2-3,5-dimethylpyrazol-1-yl-6-methylpyrimidin-4-amine,n-cyclohexyl-n-2-3,5-dimethyl-pyrazol-1-yl-6-methyl-4-pyrimidinamine,d00dif,cyppa hplc , solid,2-3,5-dimethyl-1h-pyrazole-1-yl-4-methyl-6-cyclohexylamino pyrimidine,n-cyclohexyl-2-3,5-dimethyl-1h-pyrazol-1-yl-6-methyl-4-pyrimidinamine,n-cyclohexyl-n-2-3,5-dimethyl-pyrazol-1-yl-6-methyl-4-pyrimidinamine
909822
N-cyclohexyl-2-(3,5-dimethylpyrazol-1-yl)-6-methylpyrimidin-4-amine
CC1=CC(=NC(=N1)N2C(=CC(=N2)C)C)NC3CCCCC3

Activator of KCa2.2 and KCa2.3 channels

Activator of small conductance Ca2+-activated K+ channels that displays selectivity for KCa2.2 (SK2) and KCa2.3 (SK3) channels (EC50 values are 5.6 and 14 μM for KCa2.3 and KCa2.2 respectively). Displays no activity at KCa2.1 (SK1) and KCa3.1 (IK) channels.
Quantity 10mg
Formula Weight 285.39
Percent Purity >99%
Chemical Name or Material CyPPA

Recommended Storage : Store at +4°C