Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyproheptadine Hydrochloride, USP, 98.5-100.5%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
41354-29-4
C42H50Cl2N2O3
701.77
ZEAUHIZSRUAMQG-UHFFFAOYSA-N
dihydrogen bis(1-methyl-4-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-ylidene}piperidine) trihydrate dichloride
[H+].[H+].O.O.O.[Cl-].[Cl-].CN1CCC(CC1)=C1C2=CC=CC=C2C=CC2=CC=CC=C12.CN1CCC(CC1)=C1C2=CC=CC=C2C=CC2=CC=CC=C12
41354-29-4
C42H50Cl2N2O3
701.77
ZEAUHIZSRUAMQG-UHFFFAOYSA-N
dihydrogen bis(1-methyl-4-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-ylidene}piperidine) trihydrate dichloride
[H+].[H+].O.O.O.[Cl-].[Cl-].CN1CCC(CC1)=C1C2=CC=CC=C2C=CC2=CC=CC=C12.CN1CCC(CC1)=C1C2=CC=CC=C2C=CC2=CC=CC=C12

C1968, 41354-29-4, C21H21N•HCl•11/2H2O

Spectrum™ Cyproheptadine Hydrochloride, USP , a first-generation antihistamine, is used to treat allergic reactions. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
UN Number UN2811
Ignition Residue 0.1%
Percent Purity 98.5 to 100.5%
Packaging Amber Glass Bottle
Quantity 1 g