Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyr61/CCN1 Mouse anti-Human, FITC, Clone: 3A7.1B8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236491F

Catalog No. NBP236491F


Add to cart

Description

Description

CD98 Monoclonal antibody specifically detects CD98 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

CD98
1C11.7E3
FITC
PBS with 0.05% Sodium Azide
4F2, 4F2hc, 4T2HC, antigen identified by monoclonal antibodies 4F2, TRA1.10, TROP4, and T43,4F2HC, CD98, CD98 antigen, CD98 heavy chain, CD98HC, heavy chain, Lymphocyte activation antigen 4F2 large subunit, MDU1antigen defined by monoclonal 4F2, heavy chain, monoclonal 44D7, NACAE4F2 cell-surface antigen heavy chain, solute carrier family 3 (activators of dibasic and neutral amino acidtransport), member 2,4F2 heavy chain antigen
Mouse
IgG2b Kappa
0.1 ml
Biologically Active Proteins
Primary
6520
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
1.0 mg/mL
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
SLC3A2
Partial recominant human CD98 (between amino acids 200-400) [UniProt P08195]
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only