Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyr61/CCN1 Mouse anti-Human, Janelia Fluor 549, Clone: 3H3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP104285JF549

Catalog No. NBP14285549

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Cyr61/CCN1 Monoclonal antibody specifically detects Cyr61/CCN1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Cyr61/CCN1
3H3
Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
CYR61
Recombinant human CYR61 (168-242aa) purified from E. coli (NP_001545).
Protein G purified
Biologically Active Proteins, GPCR
Primary
3491
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CCN family member 1, CCN1IGF-binding protein 10, Cysteine-rich angiogenic inducer 61, cysteine-rich, angiogenic inducer, 61, GIG1cysteine-rich heparin-binding protein 61, IBP-10, IGFBP-10, IGFBP10cysteine-rich, anigogenic inducer, 61, Insulin-like growth factor-binding protein 10, protein CYR61, Protein GIG1
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Anti-human CYR61 mAb, clone 3H3, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human CYR61 protein.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit