Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYRPTIC/CFC1 Antibody (MM0205-8D12), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212209AF405
Description
CYRPTIC/CFC1 Monoclonal specifically detects CYRPTIC/CFC1 in Human samples. It is validated for Western Blot.Specifications
CYRPTIC/CFC1 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CFC1 | |
Human recombinant CYRPTIC/CFC1 | |
0.1 mL | |
55997 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot | |
MM0205-8D12 | |
Western Blot | |
CFC1B, cripto, FRL-1, cryptic family 1, Cryptic family protein 1, cryptic protein, CRYPTICFLJ77897, HTX2MGC133213 | |
Mouse | |
Protein G purified | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction