Learn More
CYRPTIC/CFC1 Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0205-8D12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212209AF350
Catalog No. NP212209A30
Add to cart
Description
Description
CYRPTIC/CFC1 Monoclonal antibody specifically detects CYRPTIC/CFC1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
CYRPTIC/CFC1
MM0205-8D12
Western Blot
CFC1B, cripto, FRL-1, cryptic family 1, Cryptic family protein 1, cryptic protein, CRYPTICFLJ77897, HTX2MGC133213
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
CFC1
Human CFC1
0.1 mL
Primary
55997