Learn More
CYRPTIC/CFC1 Rat anti-Mouse, Alexa Fluor™ 750, Clone: RM0198-1M10, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212210AF750
Catalog No. NP212210A70
Add to cart
Description
Description
CYRPTIC/CFC1 Monoclonal antibody specifically detects CYRPTIC/CFC1 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
CYRPTIC/CFC1
RM0198-1M10
Western Blot
CFC1B, cripto, FRL-1, cryptic family 1, Cryptic family protein 1, cryptic protein, CRYPTICFLJ77897, HTX2MGC133213
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CFC1
Mouse CFC1
0.1 mL
Primary
55997