Learn More
[Cys3,6, Tyr8, Pro10] Substance P, Alfa Aesar™
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar J66549LB0
 View more versions of this product
Catalog No. AAJ66549LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1295.56
5mg
Arg-Pro-Cys-Pro-Gln-Cys-Phe-Tyr-Gly-Pro-Met-NH2, (Disulfide bridge: Cys3-Cys6)
-30° to -10°C
HPLC
Solid
RUO
[Cys3,6, Tyr8, Pro10] Substance P