Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ cysH (Escherichia coli) Recombinant Protein
Used for SDS-PAGE
Supplier: Abnova™ P5004
Description
Sequence: MGSSHHHHHHSSGLVPRGSHMSKLDLNALNELPKVDRILALAETNAELEKLDAEGRVAWALDNLPGEYVLSSSFGIQAAVSLHLVNQIRPDIPVILTDTGYLFPETYRFIDELTDKLKLNLKVYRATESAAWQEARYGKLWEQGVEGIEKYNDINKVEPMNRALKELNAQTWFAGLRREQSGSRANLPVLAIQRGVFKVLPIIDWDNRTIYQYLQKHGLKYHPLWDEGYLSVGDTHTTRKWEPGMAEEETRFFGLKRECGLHEGSpecifications
NP_417242 | |
SDS-PAGE | |
947230 | |
cysH (Escherichia coli) Recombinant Protein | |
Conventional Chromatography | |
100 ug | |
Store at 4°C for 1-2 weeks. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. | |
ECK2757/JW2732 | |
cysH | |
His | |
Liquid |
0.5 mg/mL | |
Liquid | |
30.1kDa | |
Escherichia coli expression system | |
Loading 3 ug protein in 15% SDS-PAGE | |
MGSSHHHHHHSSGLVPRGSHMSKLDLNALNELPKVDRILALAETNAELEKLDAEGRVAWALDNLPGEYVLSSSFGIQAAVSLHLVNQIRPDIPVILTDTGYLFPETYRFIDELTDKLKLNLKVYRATESAAWQEARYGKLWEQGVEGIEKYNDINKVEPMNRALKELNAQTWFAGLRREQSGSRANLPVLAIQRGVFKVLPIIDWDNRTIYQYLQKHGLKYHPLWDEGYLSVGDTHTTRKWEPGMAEEETRFFGLKRECGLHEG | |
RUO | |
cysH | |
E. coli | |
Escherichia coli expression system | |
>90% by SDS-PAGE |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction