Learn More
Tocris Bioscience™ Cystamine dihydrochloride Available on GSA/VA Contract for Federal Government customers only.
Transglutaminase inhibitor; neuroprotective
Manufacturer:  Tocris Bioscience™ 4981/50
Catalog No. 49-815-0R
Add to cart
This item is not returnable. View return policy
Description
Description
Transglutaminase inhibitor. Attenuates cell death induced by 3-nitropropionic acid (Cat. No. 4849) in Huntington's disease (HD) striatal cells. Also decreases aggregated and cross-linked huntingtin in transfected cells, and increases brain levels of BDNF. Shown to extend survival and improve motor performance in transgenic HD mice. Neuroprotective.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Cystamine dihydrochloride
Cystamine dihydrochloride
225.2
50mg