Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cystamine dihydrochloride, 97+%, Alfa Aesar™

Click to view available options
Quantity:
25g
100g
56-17-7
C4H14Cl2N2S2
225.19
MFCD00012905
YUFRRMZSSPQMOS-UHFFFAOYSA-N
cystamine dihydrochloride,cystamin dihydrochloride,ethanamine, 2,2'-dithiobis-, dihydrochloride,usaf cb-34,2,2'-disulfanediyldiethanamine dihydrochloride,2-aminoethyl disulfide dihydrochloride,bis 2-aminoethyl disulfide dihydrochloride,cystamine 2hcl,2,2'-dithiodi ethylammonium dichloride,decarboxycystine dihydrochloride
5941
2-(2-aminoethyldisulfanyl)ethanamine;dihydrochloride
C(CSSCCN)N.Cl.Cl
56-17-7
C4H14Cl2N2S2
225.19
MFCD00012905
YUFRRMZSSPQMOS-UHFFFAOYSA-N
cystamine dihydrochloride,cystamin dihydrochloride,ethanamine, 2,2'-dithiobis-, dihydrochloride,usaf cb-34,2,2'-disulfanediyldiethanamine dihydrochloride,2-aminoethyl disulfide dihydrochloride,bis 2-aminoethyl disulfide dihydrochloride,cystamine 2hcl,2,2'-dithiodi ethylammonium dichloride,decarboxycystine dihydrochloride
5941
2-(2-aminoethyldisulfanyl)ethanamine;dihydrochloride
C(CSSCCN)N.Cl.Cl
Applications
Cystamine dihydrochloride acts as an anti-infective agent, which is used in the treatment of urinary tract infections. It is also used as a radiation-protective agent that interferes with sulfhydryl enzymes. Further, it serves as a heparin antagonist and sulfhydryl modifying reagent. In addition to this, it is used as an inhibitor of TGase.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
Melting Point 217°C to 220°C
Quantity 100g
Odor Odorless
Assay Percent Range 98+%
Linear Formula (H2NCH2CH2S)2·2HCl
Beilstein 3616850
Merck Index 14,2776
Formula Weight 225.2
Percent Purity ≥97%
Chemical Name or Material Cystamine dihydrochloride

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P280f
Wear protective clothing.

Warning

RTECSNumber : KR7260000

TSCA : TSCA

storageNote1 : Hygroscopic