Learn More
Cystamine dihydrochloride, 97+%, Thermo Scientific Chemicals
Description
Cystamine dihydrochloride acts as an anti-infective agent, which is used in the treatment of urinary tract infections. It is also used as a radiation-protective agent that interferes with sulfhydryl enzymes. Further, it serves as a heparin antagonist and sulfhydryl modifying reagent. In addition to this, it is used as an inhibitor of TGase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Cystamine dihydrochloride acts as an anti-infective agent, which is used in the treatment of urinary tract infections. It is also used as a radiation-protective agent that interferes with sulfhydryl enzymes. Further, it serves as a heparin antagonist and sulfhydryl modifying reagent. In addition to this, it is used as an inhibitor of TGase.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
Specifications
Melting Point | 217°C to 220°C |
Assay Percent Range | 98+% |
Linear Formula | (H2NCH2CH2S)2·2HCl |
Quantity | 25 g |
Beilstein | 3616850 |
Sensitivity | Hygroscopic |
Merck Index | 14,2776 |
Solubility Information | Soluble in water. |
Formula Weight | 225.2 |
Percent Purity | ≥97% |
Show More |
Safety and Handling
Hazard Category | H302-H315-H319-H335-H500 |
Hazard Statement | GHS H Statement H302 Harmful if swallowed. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
EINECSNumber | 200-260-7 |
RTECSNumber | KR7260000 |
TSCA | Yes |
storageNote1 | Hygroscopic |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.