Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cystatin A Mouse anti-Human, Clone: CPTC-CSTA-1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30778120UG

 View more versions of this product

Catalog No. NB123036


Add to cart

Description

Description

Cystatin A Monoclonal antibody specifically detects Cystatin A in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications

Specifications

Cystatin A
CPTC-CSTA-1
Unconjugated
10 mM PBS with 0.05% BSA
cystatin A (stefin A), cystatin AS, cystatin-A, STF1Cystatin-AS, STFAStefin-A
Recombinant full-length human Cystatin A protein
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
Western Blot, Immunohistochemistry (Paraffin)
0.2 mg/ml
Western Blot 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Purified
Mouse
IgG2c Kappa
20 μg
Cancer, Tumor Suppressors
Primary
1475
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit