Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cystatin C Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI2C8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270472JF549

Catalog No. N270472J549


Add to cart

Description

Description

Cystatin C Monoclonal antibody specifically detects Cystatin C in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

Cystatin C
OTI2C8
Immunohistochemistry
Affinity Purified, Conjugated
CST3
Full length recombinant protein of human Cystatin C (NP_000090) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
ARMD11, bA218C14.4 (cystatin C), cystatin 3, cystatin C, cystatin C (amyloid angiopathy and cerebral hemorrhage), cystatin-3, cystatin-C, Gamma-trace, MGC117328, Neuroendocrine basic polypeptide, post-gamma-globulin
Mouse
IgG1
0.1 mL
Primary
1471.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit