Learn More
Cystatin E/M/CST6 Rat anti-Mouse, Alexa Fluor 405, Clone: RM0217-6F49, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP212252AF405
Catalog No. NB212252405
Add to cart
Description
Description
Cystatin E/M/CST6 Monoclonal specifically detects Cystatin E/M/CST6 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Cystatin E/M/CST6
RM0217-6F49
Western Blot
Purified
CST6
Mouse CST6
Protein G purified
Cancer
Primary
1474
Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
cystatin 6, cystatin E/M, cystatin M, cystatin M/E, Cystatin-6, Cystatin-E, cystatin-M, cysteine proteinase inhibitor
Rat
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
Mouse