Learn More
Cystatin SA Antibody (OTI1D6), DyLight 350, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270466UV
Catalog No. NBP270466UV
Add to Cart
Description
Description
Cystatin SA Monoclonal specifically detects Cystatin SA in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
Cystatin SA
Monoclonal
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
CST2
Full length recombinant protein of human Cystatin SA (NP_001313) produced in HEK293T cells.
0.1 mL
1470.0
Store at 4°C in the dark.
Western Blot, Immunohistochemistry
OTI1D6
Western Blot, Immunohistochemistry
cystatin 2, cystatin S5, cystatin SA, Cystatin-2, Cystatin-S5, cystatin-SA, cysteine-proteinase inhibitor, MGC71924, salivary cysteine (thiol) protease inhibitor
Mouse
Affinity Purified
Primary
Human
IgG1
Product Suggestions
Product Suggestions