Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin SN Antibody (MM0234-5N51) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212255
Description
Cystatin SN Monoclonal specifically detects Cystatin SN in Human samples. It is validated for Western Blot.Specifications
Cystatin SN | |
Monoclonal | |
LYOPH | |
Western Blot 1:500-1:1000 | |
P01037 | |
CST1 | |
Human recombinant Cystatin SN | |
0.1 mg | |
Proteases & Other Enzymes | |
1469 | |
Human | |
Purified |
Western Blot | |
MM0234-5N51 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative | |
cystain-SA-I, cystatin 1, cystatin SA-I, cystatin SN, cystatin-1, cystatin-SN, Salivary cystatin-SA-1, type 2 family | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Reconstitute with sterilized PBS to a final concentration of 0.5 mg/ml. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction