Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin SN Antibody (MM0234-5N51), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212255AF594
Description
Cystatin SN Monoclonal specifically detects Cystatin SN in Human samples. It is validated for Western Blot.Specifications
Cystatin SN | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
CST1 | |
Human recombinant Cystatin SN | |
Proteases & Other Enzymes | |
1469 | |
Store at 4°C in the dark. |
Western Blot | |
MM0234-5N51 | |
Western Blot | |
cystain-SA-I, cystatin 1, cystatin SA-I, cystatin SN, cystatin-1, cystatin-SN, Salivary cystatin-SA-1, type 2 family | |
Mouse | |
0.1 mL | |
Primary | |
Human | |
IgG2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction