Learn More
Cystatin/Stefin Homolog Rat anti-Mouse, Alexa Fluor 532, Clone: RM0218-8D33, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212412AF532
Catalog No. NP212412A32
Add to cart
Description
Description
Cystatin/Stefin Homolog Monoclonal antibody specifically detects Cystatin/Stefin Homolog in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Cystatin/Stefin Homolog
RM0218-8D33
Western Blot
CSTA
Mouse recombinant Cystatin H (Cystatin/Stefin Homolog)
0.1 mL
Primary
1475
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse