Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cystatin/Stefin Homolog Rat anti-Mouse, Alexa Fluor 750, Clone: RM0218-8D33, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212412AF750

Catalog No. NP212412A70


Add to cart

Description

Description

Cystatin/Stefin Homolog Monoclonal antibody specifically detects Cystatin/Stefin Homolog in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Cystatin/Stefin Homolog
RM0218-8D33
Western Blot
Conjugated, Purified
Rat
IgG2
0.1 mL
Primary
1475
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CSTA
Mouse recombinant Cystatin H (Cystatin/Stefin Homolog)
Protein G purified
Store at 4°C in the dark.
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit