Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cystatin/Stefin Homolog Antibody (RM0218-8D33) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
$204.00 - $542.50
Specifications
Antigen | Cystatin/Stefin Homolog |
---|---|
Clone | RM0218-8D33 |
Concentration | LYOPH |
Applications | Blocking Assay, Western Blot |
Classification | Monoclonal |
Description
Cystatin/Stefin Homolog Monoclonal specifically detects Cystatin/Stefin Homolog in Mouse samples. It is validated for Western Blot, Block/Neutralize.Specifications
Cystatin/Stefin Homolog | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
XP_148650 | |
CSTA | |
Primary |
RM0218-8D33 | |
Blocking Assay, Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
1475 | |
IgG2 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title