Learn More
BioVision IncSupplier Diversity Partner Cysteine Assay Kit (Fluorometric): Cysteine Assay Kit (Fluorometric): Convenient assay to measure cysteine in serum, plasma and other biological samples. Detection limit: 10 µM. 100 Assays. HTP adaptable DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K558100
Cysteine (CYS) is a sulfhydryl-containing amino acid and an important structural and functional part of proteins. BioVision’s Cysteine Assay Kit provides a simple, sensitive, and high-throughput adaptable assay that detects physiological concentration of cysteine in a variety of biological fluids. The principle of the assay is based on the cleavage of thiol group of reduced cysteine producing a fluorophore (Ex/Em = 365/450 nm) with a stable signal, which is directly proportional to the amount of total cysteine in the sample. The assay is specific and other thiol-based amino acids do not interfere with the assay. The assay can detect as little as 10 µM of Cysteine in a variety of samples. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9771