Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cysteine Dioxygenase Type 1 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282081F
This item is not returnable.
View return policy
Description
Cysteine Dioxygenase Type 1 Polyclonal antibody specifically detects Cysteine Dioxygenase Type 1 in Human, Mouse, Rat samples. It is validated for ELISASpecifications
Cysteine Dioxygenase Type 1 | |
Polyclonal | |
PBS | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
ELISA | |
FITC | |
CDO, CDO-I, cysteine dioxygenase type 1, Cysteine dioxygenase type I, cysteine dioxygenase, type I, EC 1.13.11.20 | |
Cysteine Dioxygenase Type 1 antibody was raised against a 19 amino acid peptide near the amino terminus of human Cysteine Dioxygenase Type 1. The immunogen is located within amino acids 30 - 80 of Cysteine Dioxygenase Type 1. Amino Acid Squence: DPTEWAMYAKFDQYRYTRN | |
0.1 mL | |
Lipid and Metabolism | |
1036 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction