Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cysteine Hydrochloride, Monohydrate, USP, 98.5-101.5%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7048-04-6
C3H10ClNO3S
175.63
QIJRTFXNRTXDIP-JIZZDEOASA-N
hydrogen (2R)-2-amino-3-sulfanylpropanoic acid hydrate chloride
[H+].O.[Cl-].N[C@@H](CS)C(O)=O
7048-04-6
C3H10ClNO3S
175.63
QIJRTFXNRTXDIP-JIZZDEOASA-N
hydrogen (2R)-2-amino-3-sulfanylpropanoic acid hydrate chloride
[H+].O.[Cl-].N[C@@H](CS)C(O)=O

C1473, 7048-04-6, C3H7NO2S•HCl•H2O

Spectrum™ Cysteine Hydrochloride, Monohydrate, USP is a semi-essential amino acid. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Ignition Residue 0.4%
Iron (Fe) 30 ppm
Percent Purity 98.5 to 101.5%
Loss on Drying 8.0-12%
Packaging Poly Bottle
Specific Rotation +5.7° to +6.8°
Quantity 500 g