Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Cystic Fibrosis Transmembrane Regulator (CFTR) Ab-4, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence and immunoprecipitation experiments.

$248.00 - $1131.00

Specifications

Antigen Cystic Fibrosis Transmembrane Regulator (CFTR) Ab-4
Clone M3A7
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 7
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS1249P
View Documents
Thermo Scientific™
MS1249P
Quantity 1mL Each for $1,131.00
Add to cart
 
MS1249P0
View Documents
Thermo Scientific™
MS1249P0
Purified Ab with BSA and Azide 100μL Each for $248.00
Add to cart
 
MS1249P1
View Documents
Thermo Scientific™
MS1249P1
Purified Ab with BSA and Azide 500μL Each for $668.50
Add to cart
 
MS1249P1ABX
View Documents
Thermo Scientific™
MS1249P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $619.50
Add to cart
 
MS1249PABX
View Documents
Thermo Scientific™
MS1249PABX
Ab without BSA/Azide (Frozen) 200μL Each for $946.00
Add to cart
 
MS1249R7
View Documents
Thermo Scientific™
MS1249R7
Ready-to-use for IHC (UVLP) 7mL Each for $625.50
Add to cart
 
MS1249R1
View Documents
Thermo Scientific™
MS1249R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ Cystic Fibrosis Transmembrane Regulator (CFTR) Ab-4, Mouse Monoclonal Antibody.

CFTR (cystic fibrosis transmembrane conductance regulator) is composed of two membrane-spanning domains (MSD), two nucleotide-binding domains (NBD), and an R domain. It is structurally similar to multidrug resistance (Mdr1) protein and both are members of the superfamily of ATP-binding cassette (ABC) transporters, also known as traffic ATPases, which are implicated in the movement of various substrates. The CFTR protein is a small conductance adenosine 3',5'-cyclic monophosphate (cAMP)-activated chloride ion channel found in the apical membranes of epithelia within the pancreas, airway, intestine, bile duct, sweat gland, and male genital ducts. CFTR is a valuable marker of human pancreatic duct cell development and differentiation.

Host Species: Mouse

Clone: M3A7

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 1370-1380

Immunogen: Recombinant protein encoding NBF2 domain of human CFTR

Molecular Weight: 165-170 kDa

Positive Control: Pancreas

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Immunofluorescence
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Cystic Fibrosis Transmembrane Regulator (CFTR) Ab-4
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
RUO
Recombinant protein encoding NBF2 domain of human CFTR
M3A7
Monoclonal
Murine
Primary
IgG1
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit