Learn More
Agilent BioTek Cytation 1 Cell Imaging Multi-Mode Reader, IVD
BioTek Cytation 1 cell imaging multi-mode reader combines fluorescence and high contrast brightfield imaging with conventional multi-mode detection. For In Vitro Diagnostic Use.
Supplier: Agilent BioTek CYT1FASI
Description
BioTek Cytation 1 cell imaging multi-mode reader combines fluorescence and high contrast brightfield imaging with conventional multi-mode detection. This affordable imaging reader delivers performance and economy compared to other digital microscopy systems
The BioTek Cytation 1 design provides both quantitative phenotypic cellular data with well-based quantitative data. The multi-mode detection module has high sensitivity filter-based fluorescence and a monochromator for UV-Vis absorbance. Temperature control and shaking are standard. CO2/O2 control and reagent injectors are available. Gen5 software simplifies image capture and plate reading.
Specifications
Cell Imaging Multi-Mode Microplate Reader | |
0 to 20% | |
Monochromator absorbance; Filter/dichroic fluorescence; Luminescence/filtered luminescence | |
6-well to 384-well plates | |
Xenon Flash | |
Gen5 Microplate Reader and Imager Software | |
20 in. | |
17.5 in. | |
16.5 in. | |
CE and TUV marked. RoHS compliant. |
Linear, Orbital, Double Orbital | |
0 to 19% | |
CYT1FA | |
Endpoint, kinetic, area scanning, plate mode inject/read process and well mode inject/read process | |
0.3 μm/pixel at 20X | |
Incubation to 45°C | |
50.8 cm | |
44.5 cm | |
41.9 cm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.