Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Cytidine-5'-Triphosphate Disodium Salt Dihydrate, Thermo Scientific™

Click to view available options
Quantity:
100mg
500mg
1g
Packaging:
Plastic bottle
81012-87-5
C9H16N3Na4O14P3
575.114
LDTPLTXNLOEHLN-ODQFIEKDSA-N
cytidine 5'-triphosphate sodium salt,cytidine 5'-triphosphate tetrasodium
129893565
[[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphono hydrogen phosphate;sodium
C1=CN(C(=O)N=C1N)C2C(C(C(O2)COP(=O)(O)OP(=O)(O)OP(=O)(O)O)O)O.[Na].[Na].[Na].[Na]
81012-87-5
C9H16N3Na4O14P3
575.114
LDTPLTXNLOEHLN-ODQFIEKDSA-N
cytidine 5'-triphosphate sodium salt,cytidine 5'-triphosphate tetrasodium
129893565
[[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphono hydrogen phosphate;sodium
C1=CN(C(=O)N=C1N)C2C(C(C(O2)COP(=O)(O)OP(=O)(O)OP(=O)(O)O)O)O.[Na].[Na].[Na].[Na]
Formula Weight 527.13 (anhydrous)
Physical Form Powder
Packaging Plastic bottle
Quantity 500mg
Chemical Name or Material Cytidine to 5'-Triphosphate Disodium Salt Dihydrate