Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYTIP Rabbit anti-Human, Alexa Fluor 405, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297434AF405
Description
CYTIP Polyclonal specifically detects CYTIP in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CYTIP | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 405 | |
B3-1, CASPCytohesin-associated scaffolding protein, Cbp HE, CYBRCytohesin-binding protein HE, CYTHIP, cytohesin 1 interacting protein, Cytohesin binder and regulator, cytohesin-1 interacting protein, cytohesin-interacting protein, HEPleckstrin homology Sec7 and coiled-coil domains-binding protein, pleckstrin homology, Sec7 and coiled/coil domains, binding protein, pleckstrin homology, Sec7 and coiled-coil domains, binding protein, PSCDBPcytohesin binding protein HE | |
Produced in rabbits immunized with E. coli-derived Human CYTIP fragment. | |
0.1 mL | |
Signal Transduction | |
9595 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title