Thermo Scientific™ Cyto-Plex™ Carboxylated Particles Available on GSA/VA Contract for Federal Government customers only.

Save time when measuring analytes with Thermo Scientific™ Cyto-Plex™ Carboxylated Particles, open platform particles for suspension array analysis.

Manufacturer: Thermo Scientific™ FM4CR11B

 View more versions of this product

Catalog No. 09-980-722

  Check Availability
Add to Cart

Alert

Contains sodium azide.


    Specifications

    Uniformity ≤2% CV
    Diameter (Metric) 4µm
    Surface Modification Carboxylate-Modified
    Color Red
    Additive Sodium Azide Preservative, 0.05% Tween-20
    Description Cyto-Plex Carboxylated Particles, 4µm
    Dye Type Firefli™ Fluorescent Red (635/700nm)
    Includes Material Safety Data Sheet (MSDS) and Package Insert Sheet
    Level Level 11
    Particle Density 1.06g/cm³
    Particle Size (Metric) 4µm
    Quantity 5mL
    Stability Product stable for a minimum of 12 months
    Storage Requirements Refrigerate when not in use; do not freeze. Store upright and keep bottle tightly sealed. Mix product by gentle inversion by hand or vortex mixer.
    Composition Polystyrene with copolymer grafted surface
    Contents Polystyrene microspheres in water
    Volume (Metric) 5mL

    Save time when measuring analytes with Thermo Scientific™ Cyto-Plex™ Carboxylated Particles, open platform particles for suspension array analysis.

    Suspension array analysis is a powerful technique enabling the simultaneous detection and quantitation of multiple analytes with a single sample. Using color coded microspheres with the appropriate dyes, these tests can now be carried out using commonly available flow cytometers.

    • Cyto-Plex™ microspheres consist of a highly uniform polystyrene particle with fluorescent intensities which are completely separated from each other
    • Carboxylated particles provide different sizes and levels of fluorescent intensities for analysis of multiple analytes
    • Use of a single diameter particle for all dye levels saves time by only requiring the development and optimization of one bead chemistry; multiple diameters can be combined to increase the number of analytes measured in one test
    • High-density binding sites and low non-specific binding enable coupling with a wide variety of antibodies, nucleic acids and other biomolecules
    • Microspheres have a maximum emission at 700 nm and can be excited with either 488 nm or 633 nm lasers
    • Emission can be collected in either the PE-Cy5 or APC channels
    • Since there is little or no emission in the FITC and PE channels, probes utilizing either of these dyes can be effectively used as reporters

    Recommended for:

    • Open platform microspheres for suspension array analysis in flow cytometry

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit