Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Cyto-Plex™ Carboxylated Particles Available on GSA/VA Contract for Federal Government customers only.

Save time when measuring analytes with Thermo Scientific™ Cyto-Plex™ Carboxylated Particles, open platform particles for suspension array analysis.

Manufacturer:  Thermo Scientific™ FM4CR05B

 View more versions of this product

Catalog No. 09-980-710


  Check Availability
Add to cart

Alert:

Contains sodium azide.


Specifications

Description

Save time when measuring analytes with Thermo Scientific™ Cyto-Plex™ Carboxylated Particles, open platform particles for suspension array analysis.

Suspension array analysis is a powerful technique enabling the simultaneous detection and quantitation of multiple analytes with a single sample. Using color coded microspheres with the appropriate dyes, these tests can now be carried out using commonly available flow cytometers.
  • Cyto-Plex™ microspheres consist of a highly uniform polystyrene particle with fluorescent intensities which are completely separated from each other
  • Carboxylated particles provide different sizes and levels of fluorescent intensities for analysis of multiple analytes
  • Use of a single diameter particle for all dye levels saves time by only requiring the development and optimization of one bead chemistry; multiple diameters can be combined to increase the number of analytes measured in one test
  • High-density binding sites and low non-specific binding enable coupling with a wide variety of antibodies, nucleic acids and other biomolecules
  • Microspheres have a maximum emission at 700 nm and can be excited with either 488 nm or 633 nm lasers
  • Emission can be collected in either the PE-Cy5 or APC channels
  • Since there is little or no emission in the FITC and PE channels, probes utilizing either of these dyes can be effectively used as reporters

Recommended for:

  • Open platform microspheres for suspension array analysis in flow cytometry
Description & Specifications

Specifications

Sodium Azide Preservative, 0.05% Tween-20
Polystyrene with copolymer grafted surface
Cyto-Plex Carboxylated Particles, 4µm
Firefli? Fluorescent Red (635/700nm)
Level 5
4µm
Product stable for a minimum of 12 months
Carboxylate-Modified
Red
Polystyrene microspheres in water
4µm
Material Safety Data Sheet (MSDS) and Package Insert Sheet
1.06g/cm³
5mL
Refrigerate when not in use; do not freeze. Store upright and keep bottle tightly sealed. Mix product by gentle inversion by hand or vortex mixer.
≤2% CV
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit