Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Cyto-Plex™ Carboxylated Particles
Save time when measuring analytes with Thermo Scientific™ Cyto-Plex™ Carboxylated Particles, open platform particles for suspension array analysis.
Supplier: Thermo Scientific™ FM4CR10
Alert:
Contains sodium azide.
Description
Save time when measuring analytes with Thermo Scientific™ Cyto-Plex™ Carboxylated Particles, open platform particles for suspension array analysis.
Suspension array analysis is a powerful technique enabling the simultaneous detection and quantitation of multiple analytes with a single sample. Using color coded microspheres with the appropriate dyes, these tests can now be carried out using commonly available flow cytometers.- Cyto-Plex™ microspheres consist of a highly uniform polystyrene particle with fluorescent intensities which are completely separated from each other
- Carboxylated particles provide different sizes and levels of fluorescent intensities for analysis of multiple analytes
- Use of a single diameter particle for all dye levels saves time by only requiring the development and optimization of one bead chemistry; multiple diameters can be combined to increase the number of analytes measured in one test
- High-density binding sites and low non-specific binding enable coupling with a wide variety of antibodies, nucleic acids and other biomolecules
- Microspheres have a maximum emission at 700 nm and can be excited with either 488 nm or 633 nm lasers
- Emission can be collected in either the PE-Cy5 or APC channels
- Since there is little or no emission in the FITC and PE channels, probes utilizing either of these dyes can be effectively used as reporters
Recommended for:
- Open platform microspheres for suspension array analysis in flow cytometry
Specifications
Sodium Azide Preservative, 0.05% Tween-20 | |
Polystyrene microspheres in water | |
Red | |
Carboxylated Particle | |
Material Safety Data Sheet (MSDS) and Package Insert Sheet | |
1.06g/cm³ | |
Product stable for a minimum of 12 months | |
≤2% CV |
Refrigerate when not in use; do not freeze. Store upright and keep bottle tightly sealed. Mix product by gentle inversion by hand or vortex mixer. | |
Cyto-Plex Carboxylated Particles, 4µm | |
Polystyrene with Copolymer Grafted Surface | |
Firefli™ Fluorescent Red (635/700nm) | |
Level 10 | |
1 mL | |
Carboxylate-Modified | |
4 μm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction