Learn More
Alfa Aesar™ Cytochalasin A
Manufacturer:  Alfa Aesar™ J60280LB0
 View more versions of this product
Catalog No. AAJ60280LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
5mg
2°C to 8°C
Cytochalasin A
14110-64-6
477.60
191°
White
Powder
An anti-cytoskeletal compound. Inhibits glucose transport, actin polymerization and blocks the formation of microtubules. Inhibits cell divisiion. Inhibits HIV-1 protease
C29H35NO5
Light Sensitive
Soluble in DMF, DMSO, ethanol and acetone. Insoluble in water.