Learn More
BioVision IncSupplier Diversity Partner Cytochrome Oxidase Activity Colorimetric Assay Kit: Cytochrome Oxidase Activity Colorimetric Assay Kit: Rapidly measures cytochrome oxidase activity from purified mitochondria or cell/tissue extracts. Simple & HTS. 100 assays DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K287100
Cytochrome Oxidase Activity Colorimetric Assay Kit: Cytochrome c Oxidase (EC 1.9.3.1) or Complex IV is the fourth complex of the Electron Transport Chain located in the mitochondrial (or bacterial) membrane. It provides energy to the cell by coupling electron transport through the Cytochrome c chain with the process of oxidative phosphorylation. It receives an electron from each of the four Cytochrome c molecules, and transfers it to one oxygen molecule, converting it into two molecules of water. In this process, it also binds to four proton molecules and translocates them across the membrane to establish electrochemical gradient. Cytochrome Oxidase Activity Assay Kit is simple, fast and high-throughput adaptable. This assay kit can be used for purified mitochondria or tissue extracts containing mitochondria. The activity of the enzyme is determined colorimetrically by following the oxidation of reduced Cytochrome c as an absorbance decrease at 550 nm.
Catalog No. 50-111-9668