Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Cytochrome b-245/p22phox Mouse anti-Human, Clone: CS9, MilliporeSigma™

Mouse Monoclonal Antibody

Specifications

Antigen Cytochrome b-245/p22phox
Clone CS9
Applications Immunoprecipitation, Inhibition Assays, Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABS2191100
View Documents
MilliporeSigma
MABS2191100UG
100 μg
Each for $387.00
N/A
MABS219125
View Documents
MilliporeSigma
MABS219125UG
25 μg
Each for $391.00
N/A
Description

Description

Cytochrome b-245/p22phox Monoclonal antibody specifically detects Cytochrome b-245/p22phox in Human samples. It is validated for Immunoprecipitation, Inhibition, Western Blotting.
Specifications

Specifications

Cytochrome b-245/p22phox
Immunoprecipitation, Inhibition Assays, Western Blot
Purified
RUO
Purified mouse monoclonal antibody IgG1 in PBS without azide.
Monoclonal
Protein G purified
CS9
Unconjugated
Mouse
Human
Partially purified human neutrophil flavocytochrome b.
Primary
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit