Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Cytochrome c Ab-3, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in ELISA, flow cytometry and immunoprecipitation experiments with Thermo Scientific™ Cytochrome c Ab-3, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS1273P1

 View more versions of this product

Catalog No. MS1273P1

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Obtain accurate, reproducible results in ELISA, flow cytometry and immunoprecipitation experiments with Thermo Scientific™ Cytochrome c Ab-3, Mouse Monoclonal Antibody.

Cytochrome c is an electron transporting protein that resides within the inter-membrane space of the mitochondria, where it plays a critical role in the process of oxidative phosphorylation and production of cellular ATP. An increasing amount of interest has been directed toward the role which cytocrome c has been demonstrated to play in apoptotic processes. Following exposure to apoptotic stimuli, cytochrome c is rapidly released from the mitochondria into the cytosol, an event which may be required for the completion of apoptosis in some systems. Cytosolic cytochrome c functions in the activation of caspase-3, and ICE family molecule that is a key effector of apoptosis.

Host Species: Mouse

Clone: CTC03; same as 2B5

Isotype: IgG2a/κ

Species Reactivity: Human, Cow, Horse and Dog. Others not known.

Epitope: Not determined

Immunogen: Equine cytochrome c

Molecular Weight: 15 kDa

Positive Control: HeLa cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting (Not suitable)
  • ELISA
  • Flow Cytometry
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Cytochrome c Ab-3
Monoclonal
200μg/mL
Concentrated
Equine cytochrome c
500μL
Cancer and Tumor Biology
Primary
Bovine, Canine, Equine, Human
Flow Cytometry, Immunoprecipitation, Western Blot
CTC03; same as 2B5
Unconjugated
Murine
IgG2a kappa
RUO
Primary
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit