Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome c Mouse anti-Human, Mouse, Rat, Amphibian, Canine, Drosophila, Equine, Pigeon, Clone: rCYCS/1010, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30761820UG
Description
Cytochrome c Monoclonal specifically detects Cytochrome c in Human, Mouse, Rat, Amphibian, Canine, Drosophila, Equine, Pigeon samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin.Specifications
Cytochrome c | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml | |
CYCHCS, cytochrome c, cytochrome c, somatic, THC4 | |
Synthetic peptides corresponding to amino acid 1-80, 81-104 and 66-104 of pigeon Cytochrome c | |
20 μg | |
Apoptosis, Cellular Markers, Cholesterol Metabolism, Core ESC Like Genes, Lipid and Metabolism, Mitochondrial Markers, Stem Cell Markers | |
54205 | |
Store at 4°C. | |
IgG1 κ |
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
rCYCS/1010 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Amphibian, Canine, Drosophila, Equine, Pigeon | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction