Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome C Oxidase subunit 6c Mouse anti-Human, DyLight™ 755, Clone: OTI4B11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270430IR

Catalog No. NBP270430IR


Add to cart

Description

Description

Cytochrome C Oxidase subunit 6c Monoclonal antibody specifically detects Cytochrome C Oxidase subunit 6c in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Cytochrome C Oxidase subunit 6c
OTI4B11
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
COX6C
Full length recombinant protein of human Cytochrome C Oxidase subunit 6c (NP_004365) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
Cytochrome c oxidase polypeptide VIc, cytochrome c oxidase subunit 6C, cytochrome c oxidase subunit VIc, cytochrome c oxidase subunit VIc preprotein
Mouse
IgG2b
0.1 mL
Primary
1345.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit