Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome C Oxidase subunit 6c Mouse anti-Human, FITC, Clone: OTI4B11, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270430F
Description
Cytochrome C Oxidase subunit 6c Monoclonal antibody specifically detects Cytochrome C Oxidase subunit 6c in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry, Immunofluorescence.Specifications
Cytochrome C Oxidase subunit 6c | |
OTI4B11 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Affinity Purified, Conjugated | |
COX6C | |
Full length recombinant protein of human Cytochrome C Oxidase subunit 6c (NP_004365) produced in HEK293T cells. | |
Immunogen affinity purified | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
FITC | |
PBS with 0.05% Sodium Azide | |
Cytochrome c oxidase polypeptide VIc, cytochrome c oxidase subunit 6C, cytochrome c oxidase subunit VIc, cytochrome c oxidase subunit VIc preprotein | |
Mouse | |
IgG2b | |
0.1 mL | |
Primary | |
1345.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title