Learn More
Cytochrome P450 17A1 Antibody (OTI3F11), DyLight 350, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270544UV
Catalog No. NBP270544UV
Add to Cart
Description
Description
Cytochrome P450 17A1 Monoclonal specifically detects Cytochrome P450 17A1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
Specifications
Cytochrome P450 17A1
Monoclonal
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
CYP17A1
Full length recombinant protein of human Cytochrome P450 17A1 (NP_000093) produced in HEK293T cells.
0.1 mL
Primary
Human
IgG1
Western Blot, Flow Cytometry, Immunohistochemistry
OTI3F11
Western Blot, Flow Cytometry, Immunohistochemistry
CPT7, cyp17, CYP17A1, cytochrome p450 XVIIA1, cytochrome P450, family 17, subfamily A, polypeptide 1, cytochrome P450, subfamily XVII (steroid 17-alpha-hydroxylase), adrenalhyperplasia, Cytochrome P450c17, Cytochrome P450-C17, EC 1.14.99, P450C17, P450C17CYPXVII, S17AH
Mouse
Affinity Purified
Lipid and Metabolism
1586.0
Store at 4°C in the dark.
Recommendations
Recommendations