Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 17A1 Mouse anti-Human, PE/Cy5.5, Clone: OTI3F11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270544PECY55

Catalog No. N270544PEC5


Add to cart

Description

Description

Cytochrome P450 17A1 Monoclonal antibody specifically detects Cytochrome P450 17A1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Cytochrome P450 17A1
OTI3F11
Flow Cytometry
Affinity Purified, Conjugated
CYP17A1
Full length recombinant protein of human Cytochrome P450 17A1 (NP_000093) produced in HEK293T cells.
Immunogen affinity purified
Lipid and Metabolism
Primary
1586.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
CPT7, cyp17, CYP17A1, cytochrome p450 XVIIA1, cytochrome P450, family 17, subfamily A, polypeptide 1, cytochrome P450, subfamily XVII (steroid 17-alpha-hydroxylase), adrenalhyperplasia, Cytochrome P450c17, Cytochrome P450-C17, EC 1.14.99, P450C17, P450C17CYPXVII, S17AH
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit