Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome P450 1A2 Mouse anti-Human, HRP, Clone: OTI8F1, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270518H
Description
Cytochrome P450 1A2 Monoclonal antibody specifically detects Cytochrome P450 1A2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Cytochrome P450 1A2 | |
OTI8F1 | |
Western Blot, Immunohistochemistry | |
Affinity Purified, Conjugated | |
CYP1A2 | |
Full length recombinant protein of human Cytochrome P450 1A2 (NP_000752) produced in HEK293T cells. | |
Immunogen affinity purified | |
Cancer, Lipid and Metabolism | |
Primary | |
1544.0 |
Immunohistochemistry, Western Blot | |
HRP | |
PBS with No Preservative | |
aryl hydrocarbon hydroxylase, CP12, CYPIA2, Cytochrome P(3)450, cytochrome P450 1A2, Cytochrome P450 4, cytochrome P450, family 1, subfamily A, polypeptide 2, cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 2, Cytochrome P450-P3, dioxin-inducible P3-450, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, P3-450, P450 form 4, P450(PA), xenobiotic monooxygenase | |
Mouse | |
IgG2a | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title