Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome P450 1A2 Mouse anti-Human, Rat, Clone: 3B8C1, Novus Biologicals™
Mouse Monoclonal Antibody
$240.50 - $458.00
Specifications
Antigen | Cytochrome P450 1A2 |
---|---|
Clone | 3B8C1 |
Dilution | Western Blot 1 - 2 ug/ml, Immunohistochemistry 1 - 5 ug/ml |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
Cytochrome P450 1A2 Monoclonal antibody specifically detects Cytochrome P450 1A2 in Human, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Cytochrome P450 1A2 | |
Western Blot 1 - 2 ug/ml, Immunohistochemistry 1 - 5 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
aryl hydrocarbon hydroxylase, CP12, CYPIA2, Cytochrome P(3)450, cytochrome P450 1A2, Cytochrome P450 4, cytochrome P450, family 1, subfamily A, polypeptide 2, cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 2, Cytochrome P450-P3, dioxin-inducible P3-450, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, P3-450, P450 form 4, P450(PA), xenobiotic monooxygenase | |
Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with rat cytochrome p450 proteins and mouse myeloma Ag8563 cells. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
3B8C1 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Cancer, Lipid and Metabolism | |
PBS | |
1544 | |
IgG1 | |
Protein A or G purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title