Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 2A6 Antibody (OTI1D2) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270524

Catalog No. NBP270524


Add to Cart

Description

Description

Cytochrome P450 2A6 Monoclonal specifically detects Cytochrome P450 2A6 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Specifications

Specifications

Cytochrome P450 2A6
Monoclonal
LYOPH
Western Blot 1:500-2000, Flow Cytometry 1:100, Immunohistochemistry 1:150, CyTOF-ready
Coumarin 7-hydroxylase, CPA6, CYP2A3P450C2A, CYP2AEC 1.14.14.1, CYPIIA6, cytochrome P450 2A6, Cytochrome P450 IIA3, Cytochrome P450(I), cytochrome P450, family 2, subfamily A, polypeptide 6, cytochrome P450, subfamily IIA (phenobarbital-inducible), polypeptide 6, flavoprotein-linked monooxygenase, P450PB, xenobiotic monooxygenase
Mouse
56.3 kDa
100 μg
Primary
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Store at -20°C. Avoid freeze-thaw cycles.
IgG2b
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
OTI1D2
Unconjugated
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
CYP2A6
Full length human recombinant protein of human CYP2A6(NP_000753) produced in HEK293T cell.
Affinity Purified
Lipid and Metabolism
1548
Human, Mouse, Rat, Primate, Monkey
Lyophilized
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit