Learn More
Cytochrome P450 2A6 Antibody (OTI1D2), DyLight 488, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270524G
Catalog No. NBP270524G
Add to Cart
Description
Description
Cytochrome P450 2A6 Monoclonal specifically detects Cytochrome P450 2A6 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
Specifications
Cytochrome P450 2A6
Monoclonal
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
CYP2A6
Full length recombinant protein of human Cytochrome P450 2A6 (NP_000753) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat, Primate, Monkey
IgG2b
Western Blot, Flow Cytometry, Immunohistochemistry
OTI1D2
Western Blot, Flow Cytometry, Immunohistochemistry
Coumarin 7-hydroxylase, CPA6, CYP2A3P450C2A, CYP2AEC 1.14.14.1, CYPIIA6, cytochrome P450 2A6, Cytochrome P450 IIA3, Cytochrome P450(I), cytochrome P450, family 2, subfamily A, polypeptide 6, cytochrome P450, subfamily IIA (phenobarbital-inducible), polypeptide 6, flavoprotein-linked monooxygenase, P450PB, xenobiotic monooxygenase
Mouse
Affinity Purified
Lipid and Metabolism
1548.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions