Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome P450 2A6 Mouse anti-Human, Mouse, Rat, Monkey, HRP, Clone: OTI1D2, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270524H
Description
Cytochrome P450 2A6 Monoclonal antibody specifically detects Cytochrome P450 2A6 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Cytochrome P450 2A6 | |
OTI1D2 | |
Western Blot, Immunohistochemistry | |
Affinity Purified, Conjugated | |
CYP2A6 | |
Full length recombinant protein of human Cytochrome P450 2A6 (NP_000753) produced in HEK293T cells. | |
Immunogen affinity purified | |
Lipid and Metabolism | |
Primary | |
1548.0 |
Immunohistochemistry, Western Blot | |
HRP | |
PBS with No Preservative | |
Coumarin 7-hydroxylase, CPA6, CYP2A3P450C2A, CYP2AEC 1.14.14.1, CYPIIA6, cytochrome P450 2A6, Cytochrome P450 IIA3, Cytochrome P450(I), cytochrome P450, family 2, subfamily A, polypeptide 6, cytochrome P450, subfamily IIA (phenobarbital-inducible), polypeptide 6, flavoprotein-linked monooxygenase, P450PB, xenobiotic monooxygenase | |
Mouse | |
IgG2b | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human, Monkey, Mouse, Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title