Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 2A6 Mouse anti-Human, Mouse, Rat, Monkey, Janelia Fluor™ 549, Clone: OTI1D2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270524JF549

Catalog No. N270524J549


Add to cart

Description

Description

Cytochrome P450 2A6 Monoclonal antibody specifically detects Cytochrome P450 2A6 in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications

Specifications

Cytochrome P450 2A6
OTI1D2
Flow Cytometry, Immunohistochemistry
Affinity Purified, Conjugated
CYP2A6
Full length recombinant protein of human Cytochrome P450 2A6 (NP_000753) produced in HEK293T cells.
Immunogen affinity purified
Lipid and Metabolism
Primary
1548.0
Flow Cytometry, Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
Coumarin 7-hydroxylase, CPA6, CYP2A3P450C2A, CYP2AEC 1.14.14.1, CYPIIA6, cytochrome P450 2A6, Cytochrome P450 IIA3, Cytochrome P450(I), cytochrome P450, family 2, subfamily A, polypeptide 6, cytochrome P450, subfamily IIA (phenobarbital-inducible), polypeptide 6, flavoprotein-linked monooxygenase, P450PB, xenobiotic monooxygenase
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Monkey, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit