Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 2B6 Mouse anti-Human, Mouse, Rat, Canine, Monkey, PE/Cy5.5, Clone: OTI3D5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270526PECY55

Catalog No. N270526PEC5


Add to cart

Description

Description

Cytochrome P450 2B6 Monoclonal antibody specifically detects Cytochrome P450 2B6 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

Cytochrome P450 2B6
OTI3D5
Flow Cytometry
Affinity Purified, Conjugated
CYP2B6
Full length recombinant protein of human Cytochrome P450 2B6 (NP_000758) produced in HEK293T cells.
Immunogen affinity purified
Lipid and Metabolism
Primary
1555.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
CPB6, CYP2B, CYP2B7, CYP2B7P, CYPIIB6cytochrome P450, subfamily IIB (phenobarbital-inducible), cytochrome P450 2B6, Cytochrome P450 IIB1, cytochrome P450, family 2, subfamily B, cytochrome P450, family 2, subfamily B, polypeptide 6, cytochrome P450, subfamily IIB (phenobarbital-inducible), polypeptide 6, EC 1.14.14.1, IIB1, P450
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Monkey, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit