Learn More
Cytochrome P450 2E1 Antibody (OTI5B9), DyLight 680, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270531FR
Catalog No. NBP270531FR
Add to Cart
Description
Description
Cytochrome P450 2E1 Monoclonal specifically detects Cytochrome P450 2E1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Cytochrome P450 2E1
Monoclonal
DyLight 680
PBS with 0.05% Sodium Azide
CYP2E1
Full length recombinant protein of human Cytochrome P450 2E1 (NP_000764) produced in HEK293T cells.
0.1 mL
Primary
Human
IgG1
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI5B9
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
4-nitrophenol 2-hydroxylase, CPE1, CYP2Ecytochrome P450 2E1, CYPIIE1, cytochrome P450, family 2, subfamily E, polypeptide 1, cytochrome P450, subfamily IIE (ethanol-inducible), polypeptide 1, Cytochrome P450-J, EC 1.14.13.-, EC 1.14.13.n7, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, P450C2E, P450-J, xenobiotic monooxygenase
Mouse
Affinity Purified
Lipid and Metabolism
1571.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions